missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica gel, functionalized, diethylenetriamine tetraacetate, 0.5mmol/g, ACROS Organics™

Chemical Identifiers

CAS 7631-86-9,na
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD06660072
Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate, Functionalized, Diethylenetriamine tetraacetate, 0.5 mmol/g
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC372240010 SDS Thermo Fisher Scientific
AC372240010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC372240050 SDS Thermo Fisher Scientific
AC372240050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For solid support synthesis
Description & Specifications

Chemical Identifiers

7631-86-9,na
60.08
Silica gel, functionalized, diethylenetriamine tetraacetate, Functionalized, Diethylenetriamine tetraacetate, 0.5 mmol/g
O2Si
MFCD06660072

Specifications

7631-86-9,na
Glass bottle
Pale yellow
O2Si
MFCD06660072
Silica gel, functionalized, diethylenetriamine tetraacetate, Functionalized, Diethylenetriamine tetraacetate, 0.5 mmol/g
0.5mmol/g, exact loading on the certificate of analysis
1g
Powder
60.08
15, 8632

Safety and Handling

GHS H Statement:
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.