Please login to your online account to display your discounted pricing

Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC360181000

 View more versions of this product

Catalog No. AC360181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Quantity 100g
Color White to off-white
Particle Size Particle size: 40 to 63 Micron
Chemical Name or Material Silica gel, functionalized, Amino-3
CAS 71888-97-6
Loading 1.2mmol/g min. (molecular)
Packaging Glass bottle
Nitrogen (N) 1.68% min.
MDL Number MFCD03427643

Specifications

Chemical Name or Material Silica gel, functionalized, Amino-3
CAS 71888-97-6
MDL Number MFCD03427643