missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene, 1 mmol/g, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC372250050

 View more versions of this product

Catalog No. AC372250050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene
7631-86-9,5807-14-7
5g
60.08
15, 8632
Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g
Glass bottle
O2Si
MFCD03791338
1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel

Chemical Identifiers

7631-86-9,5807-14-7
60.08
1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel
O2Si
MFCD03791338
Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene, Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger