Please login to your online account to display your discounted pricing

Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene, 1 mmol/g, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC372250010

 View more versions of this product

Catalog No. AC372250010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 60.08
Quantity 1g
Merck Index 15, 8632
Chemical Name or Material Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene
Name Note Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g
Molecular Formula O2Si
MDL Number MFCD03791338
CAS 7631-86-9
Synonym 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
CAS 5807-14-7
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene
Name Note Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g
CAS 7631-86-9
CAS 5807-14-7
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD03791338
Synonym 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel
Accessories

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents