Please login to your online account to display your discounted pricing

Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene, 1 mmol/g, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene
Name Note Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g
CAS 7631-86-9
CAS 5807-14-7
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD03791338
Synonym 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372250050

  SDS
thermo fisher scientific
AC372250050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372250010

  SDS
thermo fisher scientific
AC372250010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.08
Quantity 5g
Merck Index 15, 8632
Health Hazard 1 Danger
MDL Number MFCD03791338
CAS 7631-86-9
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Name Note Functionalized, 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, 1.4 mmol/g
Packaging Glass bottle
CAS 5807-14-7
Synonym 1,5,7-Triazabicyclo[4,4,0]dec-5-ene, immobilized on silicagel
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula O2Si
Chemical Name or Material Silica gel, functionalized, 1,5,7-triazabicyclo[4,4,0]dec-5-ene

For solid support synthesis

Safety and Handling

Danger

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.