missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Indicating Desiccants, -6+16 mesh granules, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 44389A7

 View more versions of this product

Catalog No. AA44389A7



Specifications

Specifications

Silica gel desiccant
7631-86-9
5kg
MFCD00011232
Odorless
Hygroscopic
O=[Si]=O
60.083
CHEBI:30563
ACS
Blue granules
2.17 to 2.66g/cm3
O2Si
-6+16 Mesh
Soluble in alkalies. Insoluble in water.
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
Granular

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232
24261
dioxosilane
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H350i-H360F

EINECSNumber : 231-545-4

RTECSNumber : VV7565000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents