missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Desiccants, -6+12 mesh granules, Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 44387A1

 View more versions of this product

Catalog No. AA44387A1



Specifications

Specifications

Silica gel desiccant
7631-86-9
1kg
MFCD00011232
Odorless
Hygroscopic
O=[Si]=O
60.083
CHEBI:30563
White granules
2.17 to 2.66g/cm3
O2Si
-6+12 Mesh
Soluble in alkalies. Insoluble in water.
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
Granular

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232
24261
dioxosilane
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents