Learn More
Silica gel desiccant, -3+8 Mesh Granules, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 044386A7
Description
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Granular | |
Silica gel desiccant | |
Silica Gel | |
O2Si | |
-3+8 Mesh | |
Soluble in alkalies. Insoluble in water. | |
O=[Si]=O | |
dioxosilane | |
24261 |
White | |
7631-86-9 | |
5 kg | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Odorless | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
Hygroscopic | |
60.08 | |
CHEBI:30563 |
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
24261 | |
dioxosilane |
Safety and Handling
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only