missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Desiccants, Alfa Aesar™

Manufacturer:  Alfa Aesar 44386A1

 View more versions of this product

Catalog No. AA44386A1



Specifications

Specifications

Silica gel desiccant
7631-86-9
White
O2Si
Odorless
O=[Si]=O
60.083
CHEBI:30563
White granules
2.17 to 2.66g/cm3
1kg
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
Granular

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232
24261
dioxosilane
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

SDS Documents