missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Desiccant , Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1343-98-2
Chemical Name or Material Silica Gel Desiccant, indicating
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK4471212 Avantor Performance
447106
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK4471500 Avantor Performance
447104
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1343-98-2
Silica Gel Desiccant, indicating

Specifications

2.5kg
1343-98-2
Poly
Silica Gel Desiccant, indicating