Please login to your online account to display your discounted pricing

Silica Gel Desiccant , Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  447106

 View more versions of this product

Catalog No. MK4471212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Name Note indicating
Chemical Name or Material Silica Gel Desiccant
Packaging Poly
CAS 1343-98-2
Quantity 2.5kg

Specifications

Chemical Name or Material Silica Gel Desiccant
Name Note indicating
CAS 1343-98-2