Please login to your online account to display your discounted pricing

Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  S162212

 View more versions of this product

Catalog No. S162212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Quantity 2.5kg
CAS 7646-79-9
CAS 112926-00-8
Physical Form Solid
Mesh Size 4 to 10 Mesh
Color Blue
Molecular Formula SiO2
Suitability Pass Test
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Chemical Name or Material Silica Gel Desiccant, Indicating
Name Note 4-10 Mesh, Grade 48
MDL Number MFCD00011232
Identification Pass Test
Packaging Poly Bottle
Health Hazard 2 CAUTION!
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Silica Gel Desiccant, Indicating
Name Note 4-10 Mesh, Grade 48
CAS 112926-00-8
CAS 7646-79-9
Purity Grade Certified ACS Grade
Physical Form Solid
Molecular Formula SiO2
MDL Number MFCD00011232
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

CAUTION!

SDS & Safety Documents