Please login to your online account to display your discounted pricing

Silica Gel Desiccant (12-24 mesh, Grade 408/Certified ACS), Fisher Chemical

$2,668.54 - $2,668.54

Chemical Identifiers

Chemical Name or Material Silica Gel Desiccant
Name Note 12-24 Mesh, Grade 408
CAS 7631-86-9
Purity Grade Certified ACS Grade
Physical Form Solid
Molecular Formula SiO2
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S156-10

  SDS
fisher scientific company
FLS156-10
Poly Pail 10kg Each for $2,668.54
Description & Specifications

Specifications

Purity Grade Certified ACS Grade
Quantity 10kg
Physical Form Solid
Mesh Size 12 to 24 Mesh
Color Blue
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
CAS 7631-86-9
Name Note 12-24 Mesh, Grade 408
MDL Number MFCD00011232
Suitability Pass Test
Packaging Poly Pail
Molecular Formula SiO2
Identification Pass Test
Chemical Name or Material Silica Gel Desiccant
Health Hazard 2 CAUTION!
ChemAlert Storage Symbol Gray
Safety and Handling
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

CAUTION!