missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical™

Silica gel Grade 923 conforms to the specifications of ASTM D1319 and ASTM D2549.

Manufacturer:  Fisher Chemical S67925

 View more versions of this product

Catalog No. S67925

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Sold as a case of 3 boxes - each box contains 24 x 10 g pouches
  • Suitable for use in Fluorescent Indicator Adsorption (FIA) test
  • Conforms to the specifications of ASTM D1319 and ASTM D2549
  • Surface area within the range of 430 - 530 m2/g
  • This grade of silica gel is also available in other sizes:
    • 500 g (S679500)
    • 2.5 kg (S679212)
    • 25 g (S67925)
Specifications

Specifications

Silica Gel Desiccant
7631-86-9
0.73 GM/C.C. min.
2230°C
1710°C
25kg
0.005% max.
Inclusive between 4.5%
Fiber Drum
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
99% min.
60.083
CHEBI:30563
Inclusive between 430m2/g
100-200 Mesh
Conforms (D1319, D2549)
2.1g/cm³
White
5.5 to 7.0 (for 5% slurry)
Pass Test
O2Si
MFCD00011232
5.0% max.
O=[Si]=O
dioxosilane
24261
Solid
Chromatography

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
CHEBI:30563
O=[Si]=O
O2Si
MFCD00011232
24261
dioxosilane
Safety and Handling

Safety and Handling

SDS Documents
Product Certifications
Promotions

Promotions