missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica gel, for chromatography, 0.075-0.250 mm, 150 A, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC419280010

 View more versions of this product

Catalog No. AC419280010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

White
60.08
15, 8632
0.01% max. (on dry basis)
7631-86-9
For column chromatography
Glass bottle
275 to 375 m2/g (BET)
Very Fine Granular Powder
1kg
Nominal particle size: >60 mesh: <7.5%, <200 mesh: <10.0%
total volatiles (950°C): < 13.0%
O2Si
MFCD00011232
N2-pore volume (ml/g): 1.0 to 1.3
Silica gel

Chemical Identifiers

7631-86-9
O2Si
MFCD00011232
Very Fine Granular Powder
60.08
Silica gel, For column chromatography