missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica gel, C8-RP, 12%C, ca. 1.2mmol/g, part. size 40-63μ, Thermo Scientific Chemicals

$2,115.80 - $2,115.80

Chemical Identifiers

MDL Number MFCD00148339
Synonym Octylsilicate
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC360142500
SDS View Documents Promotion Details
Thermo Scientific Chemicals
360142500
250 g Plastic Bottle
Each for $2,115.80
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
Specifications

Chemical Identifiers

MFCD00148339
Octylsilicate

Specifications

Powder
Silica Gel
MFCD00148339
Octylsilicate
Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron
White
250 g
Plastic Bottle
40 to 63 Micron
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only