Please login to your online account to display your discounted pricing

Silica gel, C18-RP, 17%C, ca.0,8mmol/g, part. size: 40-63μ, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: fisher scientific acros  36012-0500

 View more versions of this product

Catalog No. AC360120500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 50g
Chemical Name or Material Silica gel, C18-RP, 17%C
Packaging Glass bottle
CAS 71889-02-6
Synonym Octadecylsilicate

Specifications

Chemical Name or Material Silica gel, C18-RP, 17%C
CAS 71889-02-6
Synonym Octadecylsilicate