missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel 60 SilicAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 63231-67-4
Chemical Name or Material Silica Gel 60, (63-200 Microns)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MKV1501 Avantor Performance
V15010
Poly 1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKV150P5 Avantor Performance
V15064
Poly Pail 5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

63231-67-4
Silica Gel 60, (63-200 Microns)

Specifications

Poly
63231-67-4
1kg
Silica Gel 60, (63-200 Microns)