Learn More
Silica gel 60, 0.060-0.2mm (70-230 mesh), Thermo Scientific Chemicals
$192.56 - $446.60
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL140020B
|
Thermo Scientific Chemicals
L140020B |
1000 g |
N/A
|
|
|||||
AAL140020E
|
Thermo Scientific Chemicals
L140020E |
2500 g |
N/A
|
|
|||||
Description
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and removing impurities. It is used as stationary base in column chromatography.
Solubility
Soluble in hydrofluoric acid. Insoluble in water.
Notes
Keep the container tightly sealed. Store in cool, dry conditions in a well sealed container. Incompatible with hydrogen fluoride.
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
24261 | |
dioxosilane |
Specifications
7631-86-9 | |
1000 g | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
14,8493 | |
Soluble in hydrofluoric acid. Insoluble in water. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 |
Silica Gel | |
O2Si | |
70 to 230 mesh | |
Odorless | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
Silica gel 60, 0.060 to 0.2mm |
Safety and Handling
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-545-4
RTECSNumber : VV7565000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only