Silica Gel 30 SilicAR, Macron Fine Chemicals™
macron
Manufacturer:  Avantor Performance V15213
 View more versions of this product
Catalog No. MKV1525LB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Silica Gel 30
63231-67-4
H2O3Si
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768
(100-200 Mesh) (75-150 Microns)
5lb.
Poly
O[Si](=O)O
78.098
Chemical Identifiers
63231-67-4
78.098
14768
O[Si](=O)O
H2O3Si
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
Documents