missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel 30 SilicAR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 63231-67-4
Chemical Name or Material Silica Gel 30, (100-200 Mesh) (75-150 Microns)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MKV152250 Avantor Performance
V15202
POLYSTORMOR 250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKV1525LB Avantor Performance
V15213
Poly 5lb. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

63231-67-4
Silica Gel 30, (100-200 Mesh) (75-150 Microns)

Specifications

250g
POLYSTORMOR
63231-67-4
Silica Gel 30, (100-200 Mesh) (75-150 Microns)