missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica, 32-63μm, 60Å, MP Biomedicals™
For Chromatography; Chemical Synthesis
$207.08 - $280.47
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00011232 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Chemical Identifiers
7631-86-9 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232 | |
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
7631-86-9 | |
MFCD00011232 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 | |
2g/cm3 at 20°C | |
60Å | |
500g |
O2Si | |
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 | |
Powder | |
500-600m2/g | |
White to Gray | |
Silica |
Safety and Handling
Recommended Storage : Store at Room Temperature.