Please login to your online account to display your discounted pricing

Sildenafil citrate, Tocris™

Orally active, potent PDE5 inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Sildenafil citrate
Assay Percent Range >99%
Molecular Formula C22H30N6O4S·C6H8O7
Formula Weight 666.7
Synonym 1-[[3-(4, 7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4, 3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

378450

 
r & d systems
378450
50mg Each for $844.73

378410

 
r & d systems
378410
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 666.7
Quantity 50mg
Chemical Name or Material Sildenafil citrate
CAS 171599-83-0
Assay Percent Range >99%
Molecular Formula C22H30N6O4S·C6H8O7
Recommended Storage Desiccate at Room Temperature
Synonym 1-[[3-(4, 7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4, 3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate
Safety and Handling

Desiccate at Room Temperature