Please login to your online account to display your discounted pricing

Siguazodan, Tocris


PDE3 inhibitor

Quantity 50mg

Manufacturer: r & d systems  114850

 View more versions of this product

Catalog No. 114850

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 284.32
Quantity 50mg
Chemical Name or Material Siguazodan
Assay Percent Range >99%
Molecular Formula C14H16N6O
CAS 115344-47-3
Synonym N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Siguazodan
Assay Percent Range >99%
Molecular Formula C14H16N6O
Formula Weight 284.32
Synonym N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
Accessories

Store at Room Temperature