Please login to your online account to display your discounted pricing

Siguazodan, Tocris

PDE3 inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Siguazodan
Assay Percent Range >99%
Molecular Formula C14H16N6O
Formula Weight 284.32
Synonym N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

114810

 
r & d systems
114810
10mg Each for $201.36

114850

 
r & d systems
114850
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 284.32
Quantity 10mg
Molecular Formula C14H16N6O
Chemical Name or Material Siguazodan
CAS 115344-47-3
Assay Percent Range >99%
Synonym N-Cyano-N'-methyl-N''-[4-(1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl)phenyl]guanidine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature