Learn More
Epredia™ Signature Series™ Hematoxylin
Yield results which exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
$2890.19 - $2890.19
Specifications
Description
- No filtration required
- Stains up to 2500 slides per pint
- Crisp, well-defined nuclear detail
- Slight affinity for acid mucopolysaccharide (mucin) staining
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
| 0.0 |