missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Hematoxylin

Yield results which exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.

Supplier:  Epredia™ 7212L

 Afficher plus de versions de ce produit

Numéro de catalogue. 72-12L


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • No filtration required
  • Stains up to 2500 slides per pint
  • Crisp, well-defined nuclear detail
  • Slight affinity for acid mucopolysaccharide (mucin) staining

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging
Spécifications

Spécifications

1.32 gal.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions