missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Hematoxylin 2
Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.
Supplier: Epredia™ 7231L
Description
- No filtration required
- Stains up to 2500 slides per pint
- Part of the Signature Series Staining System
- Crisp, well-defined nuclear detail chromatin delineation
Available in 5L packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
| 1.32 gal. |