missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Hematoxylin 2

Yield visually similar results to Gills 2 and 3 with this uniquely formulated product.

Supplier:  Epredia™ 7231

 View more versions of this product

Catalog No. 22-050-113


Only null left
Explore available promotions

Description

Description

  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation

Available in 5L packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

Specifications

Specifications

Hematoxylin 2
1 pt.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions