Siemens Healthineers Urinalysis Reagent Test Strips
Single-use, plastic strips with multiple reagent test areas
Manufacturer: Siemens Healthineers 2830
Description
- Ready to use and entirely disposable
- Dip and read visually for results
- Some strips can be read instrumentally on Clinitek™ 50, 100, 200, 200+, 500, Advantus, and Status Family of Urine Chemistry analyzers
Specifications
Clinitek™ 50, Clinitek™ 100, Clinitek™ 200, Clinitek™ 200+ and Clinitek™ 500 Urine chemistry analyzer | |
Yes | |
25 Pack |
Blood Urea,Nitrogen | |
Urinalysis, in-vitro diagnostic use |