Please login to your online account to display your discounted pricing

Tocris™ SIB 1893


mGlu5 antagonist and positive allosteric modulator at mGlu4

Manufacturer: tocris™  121450

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 121450R

  • / Each
 

Description & Specifications

Specifications

Inhibitors SIB 1893
Molecular Weight 195.26
Quantity 50mg
Product Type SIB 1893

A highly selective non-competitive antagonist for the metabotropic glutamate mGlu5 receptor subtype; displays an IC50 value of 0.3μM at hmGlu5, compared with > 100μM at hmGlu1b, hmGlu2, hmGlu6, hmGlu7 and hmGlu8. Centrally active upon systemic administration in vivo. Positive allosteric modulator at mGlu4 receptors.

  • Chemical Name: 2-Methyl-6-(2-phenylethenyl)pyridine
  • Formula: C14H13N
  • CAS Number: 6266-99-5
  • Solubility: Soluble to 100mM in DMSO