Please login to your online account to display your discounted pricing

Tocris™ SIB 1757


Highly selective mGlu5 antagonist

Manufacturer: tocris™  121510

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 121510

  • / Each
 

Description & Specifications

Specifications

Product Type SIB 1757
Inhibitors SIB 1757
Molecular Weight 213.24
Quantity 10mg

A highly selective antagonist for the mGlu5 metabotropic glutamate receptor subtype; displays an IC50 value of 0.4μM at hmGlu5 compared with > 30μM at hmGlu1b, hmGlu2, hmGlu4, hmGlu6, hmGlu7 and hmGlu8.

  • Chemical Name: 6-Methyl-2-(phenylazo)-3-pyridinol
  • Formula: C12H11N3O
  • CAS Number: 31993-01-8
  • Solubility: Soluble to 100mM in DMSO