missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ SIB 1757

Highly selective mGlu5 antagonist

$163.42 - $677.76

Specifications

Product Type SIB 1757
Inhibitors SIB 1757
Molecular Weight 213.24
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
121510 Tocris™
121510
10mg Each for $163.42
121550 Tocris™
121550
50mg Each for $677.76
Specifications

Description

A highly selective antagonist for the mGlu5 metabotropic glutamate receptor subtype; displays an IC50 value of 0.4μM at hmGlu5 compared with > 30μM at hmGlu1b, hmGlu2, hmGlu4, hmGlu6, hmGlu7 and hmGlu8.
  • Chemical Name: 6-Methyl-2-(phenylazo)-3-pyridinol
  • Formula: C12H11N3O
  • CAS Number: 31993-01-8
  • Solubility: Soluble to 100mM in DMSO
Description & Specifications

Specifications

SIB 1757
SIB 1757
213.24