Please login to your online account to display your discounted pricing

SIB 1553A hydrochloride, Tocris™

Subunit selective nAChR agonist

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material SIB 1553A hydrochloride
Assay Percent Range >98%
Molecular Formula C13H19NOS·HCl
Formula Weight 273.82
Synonym (±)-4-[[2-(1-Methyl-2-pyrrolidinyl)ethyl]thio]phenol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

476410

 
r & d systems
476410
10mg Each for $208.25

476450

 
r & d systems
476450
50mg Each for $867.95
Description & Specifications

Specifications

Formula Weight 273.82
Quantity 10mg
Chemical Name or Material SIB 1553A hydrochloride
Assay Percent Range >98%
Molecular Formula C13H19NOS·HCl
CAS 191611-89-9
Synonym (±)-4-[[2-(1-Methyl-2-pyrrolidinyl)ethyl]thio]phenol hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C