Please login to your online account to display your discounted pricing

SI Metric Manufacturing Sand Timers


Perfect for use in games!

Manufacturer: si metric manufacturing  16500*

 View more versions of this product

Catalog No. S93911

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For use in games, accurate event timing and experiments
Color Green
Product Type 1 min. Sand Timer

  • Virtually indestructible, with molded end caps and thick wall surrounds for accurate event timing and experiments
  • Each timer is color coded for easy identification
  • 1-, 3- and 5-minute timers available
  • Meausres: 6.3 × 2.75 in. (16 x 7cm)
  • Grades K+