Please login to your online account to display your discounted pricing

Shz 1, Tocris™

Activator of early cardiac genes in pluripotent stem cells; induces differentiation in M-PBMCs

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Shz 1
Assay Percent Range >98%
Molecular Formula C13H11BrN2O3S
Formula Weight 355.21
Synonym 2-[(5-Bromo-2-hydroxyphenyl)methylene]hydrazide benzenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

492310

 
r & d systems
RDS492310
10mg Each for $181.19

492350

 
r & d systems
RDS492350
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 355.21
Quantity 10mg
Chemical Name or Material Shz 1
Assay Percent Range >98%
Molecular Formula C13H11BrN2O3S
CAS 326886-05-9
Synonym 2-[(5-Bromo-2-hydroxyphenyl)methylene]hydrazide benzenesulfonic acid
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C