Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP) DFS Item
Shrimp Alkaline Phosphatase (SAP) is a high specific activity, heat-labile alkaline phosphatase purified from a recombinant source and originally isolated from Pandalus borealis (arctic shrimp).
Supplier:  Applied Biosystems™ 7839010KU
Catalog No. 7839010KU
Description
Description
  • 100% heat-inactivated in 5 min at 65°C
  • Significantly improved storage stability at lower temperatures
  • Very high specific activity
  • Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
  • Purified from a recombinant source
  • May be added directly to restriction enzyme digests
  • No vector purification necessary
  • Requires no supplemental zinc or other additives for activity
  • Works direct in many different buffers
  • Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Specifications
Specifications
37°C
Tested for contaminating endonucleases, exonucleases, and ribonucleases
Shrimp Alkaline Phosphatase
Shrimp Alkaline Phosphatase, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases
Shrimp Alkaline Phosphatase
0