Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ SHP-1 Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ SHP-1 Ab-1, Mouse Monoclonal Antibody.

$313.71 - $1,441.51

Specifications

Antigen SHP-1 Ab-1
Clone 1SH01; same as 11D7C8H5
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1190P

 
thermo scientific™ lab vision™
MS1190P
200μg/mL 1mL Each for $1,326.55

MS1190P0

 
thermo scientific™ lab vision™
MS1190P0
200μg/mL 100µL Each for $313.71

MS1190P1

 
thermo scientific™ lab vision™
MS1190P1
200μg/mL 500µL Each for $881.41

MS1190P1ABX

 
thermo scientific™ lab vision™
MS1190P1ABX
1mg/mL 100µL Each for $881.94

MS1190PABX

 
thermo scientific™ lab vision™
MS1190PABX
1mg/mL 200µL Each for $1,441.51
Description & Specifications

Specifications

Antigen SHP-1 Ab-1
Clone 1SH01; same as 11D7C8H5
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Immunogen Recombinant human SHP1 protein
Isotype IgG2b kappa

Provide accurate, reproducible results with the Thermo Scientific™ SHP-1 Ab-1, Mouse Monoclonal Antibody.

Shp-1 is a SH2-containing protein-tyrosine phosphatase 1 (SHP-1). SHP-1 is known to play a crucial role in the regulation of hematopoiesis and it regulates the transcriptional activity stimulated by the erythropoietin (EPO)-induced JAK/STAT and MAPK pathways and is involved in the signaling machinery responsible for erythroid differentiation and suppression of apoptosis. Furthermore, the observation that SHP-1 associates with RAFTK/Pyk2/CAK activated by β-chemokine receptor (CCR5) and SHP-1 also associates with the adaptor protein Grb2 and the Src-related kinase Syk indicates the role of SHP-1 in modulating cell migration, proliferation, and immune functions.

Host Species: Mouse

Clone: 1SH01; same as 11D7C8H5

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human SHP-1 protein

Molecular Weight: ∽68kDa

Positive Control: Jurkat or HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO