SHOWA™ Ultraflex™ II Neoprene Gloves
Highly flexible and chemical resistant
$10.83 - $133.59
Specifications
Certifications 21 CFR 170-199
Color Royal Blue
Length (English) 14 in.
Length (Metric) 35.56cm
Material Neoprene
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-168-291 SHOWA™
341409*
9 Pair for $11.33
Case of 72 Pair for $781.20
19-168-293 SHOWA™
341411
11 Dozen of 12 Pair for $128.49
Case of 6 Dozen for $748.92
19-168-292 SHOWA™
341410*
10 Pair for $10.83
Case of 72 Pair for $746.64
19-168-290 SHOWA™
341408
8 Dozen of 12 Pair for $133.59
Case of 6 Dozen for $778.68
Specifications
Description
  • Durable and abrasion resistant
  • Strong bond between neoprene coating and cotton interlock liner enhances glove flexibility and integrity
  • Wet grip
  • Withstand complete submersion
  • Length: 14 in. (35.6cm)
Description & Specifications
Specifications
21 CFR 170-199
14 in.
Neoprene
Gauntlet
Royal Blue
35.56cm
food processing, battery manufacturing, Paper pulp processing, chemical handling, Acids and Caustics, fuel handling, masonry and concrete, hexavalent Chromium protection
Two-piece interlock Cotton knit