SHOWA™ T-Flex™ Cut-Resistant Gloves
Thin, lightweight glove boasts a construction made up of only Dyneema™ cut-resistant fibers
Manufacturer:  SHOWA™ 8115XL10
 View more versions of this product
Catalog No. 29-635-2982
Description
Description
  • Outstanding ratio of cut resistance to weight
  • Good touch and tactile sensitivity
  • Suits assembly workers handling jagged rivets and sharp parts, as well as machinists replacing pinions and sharp-edged tooling parts
  • Ambidextrous
  • Thickness: 15-gauge
  • Made in the U.S.A.
Compliance
Meet FDA CR21 requirements as an under-glove for meat cutting
Specifications
Specifications
Level 3
meat cutting, construction, glove liner, recycling, bottling, Glass handling, sheet Metal handling, razor blades
X-Large, 10
Ambidextrous
Dyneema™, AlphaSan™
Light Gray
Documents