Please login to your online account to display your discounted pricing

SHOWA™ N-DEX™ Nitrile Laboratory and Industrial Gloves


Provide chemical resistance when exposed to intermittent splashing

Manufacturer: showa™  7005XXL

 View more versions of this product

Catalog No. 11-395-19E

  • / Pack of 100 Each
  • / Case of 20 Pack
 

Description & Specifications

Specifications

Certifications ASTM F 1671, 21 CFR 170-199, FDA, ISO9002
Glove Cuff Style Rolled
Hand Ambidextrous
Grade Industrial
Length (English) 9.5 in.
Length (Metric) 24cm
Material Nitrile
Powdered Yes
Size 2X-Large
Thickness 4mil
Color Blue
Disposable Yes
Coating Material Smooth