Please login to your online account to display your discounted pricing
SHOWA™ N-DEX™ Accelerator-Free Nitrile Gloves
Accelerator-free, disposable nitrile gloves are perfect for laboratory, pharmaceutical, or industrial tasks
$15.28 - $20.87
Specifications
Certifications ASTM F 1671, ASTM D 6319, NFPA 1999, 21 CFR 170-199, FDA, ISO9002, GMP
Color Hi-vis green
Material Nitrile
Disposable Yes
Glove Cuff Style Rolled
View More Specs
Products
Catalog Number Mfr. No. Coating Material Thickness Length (English) Size Price Quantity    
29-635-4043
showa™
9500PFXL10
Smooth 5mil 11 in. X-Large Pack of 50 Each for $16.05
Case of 20 Pack for $294.40
29-635-4040
showa™
9500PFS07
Smooth 5mil 11 in. Small Pack of 50 Each for $16.89
Case of 20 Pack for $309.60
29-635-4041
showa™
9500PFM08
Smooth 5mil 11 in. Medium Pack of 50 Each for $15.90
Case of 20 Pack for $291.60
29-635-4042
showa™
9500PFL09
Smooth 5mil 11 in. Large Pack of 50 Each for $16.46
Case of 20 Pack for $301.80
19-170-357B
showa™
7705PFTS
Textured fingertips 4mil 9.5 in. Small Pack of 100 Each for $20.07
Case of 20 Pack for $368.00
19-170-357D
showa™
7705PFTL
Textured fingertips 4mil 9.5 in. Large Pack of 100 Each for $20.87
Case of 20 Pack for $382.60
19-170-357A
showa™
7705PFTXS
Textured fingertips 4mil 9.5 in. X-Small Pack of 100 Each for $19.93
Case of 20 Pack for $365.40
19-170-357C
showa™
7705PFTM
Textured fingertips 4mil 9.5 in. Medium Pack of 100 Each for $20.48
Case of 20 Pack for $375.40
19-170-357E
showa™
7705PFTXL
Textured fingertips 4mil 9.5 in. X-Large Pack of 100 Each for $19.87
Case of 20 Pack for $364.20
19-130-3710
showa™
9500PFSM
Smooth 5mil 11 in. Small Pack of 50 Each for $15.28
Case of 20 Pack for $280.20
19-130-3712
showa™
9500PFLG
Smooth 5mil 11 in. Large Pack of 50 Each for $15.28
Case of 20 Pack for $280.20
19-130-3713
showa™
9500PFXL
Smooth 5mil 11 in. X-Large Pack of 50 Each for $15.28
Case of 20 Pack for $280.20
Description & Specifications
Specifications
Certifications ASTM F 1671, ASTM D 6319, NFPA 1999, 21 CFR 170-199, FDA, ISO9002, GMP
Color Hi-vis green
Material Nitrile
Disposable Yes
Glove Cuff Style Rolled
Hand Ambidextrous
Latex Free Yes
Powdered No
 Ideal for jobs requiring high visibility, quick identification.
  • Nitrile construction
  • Durable, puncture-resistant
  • Powder-free, ambidextrous
  • Available in smooth- or textured-finger styles
  • Color: High-vis Green
510K approved for medical use