Please login to your online account to display your discounted pricing
SHOWA™ Chloroflex™ Neoprene Gloves
Chlorinated for easy on and off
Manufacturer: showa™  723S07
 View more versions of this product
Catalog No. 11-395-3A
  • / Pair
  • / Case of 144 Pair
Description & Specifications
Specifications
Size 7
Length (English) 13 in.
Length (Metric) 33cm
Thickness (English) 24mil
Color Black
Certifications NFPA 1992
Glove Cuff Style Pinked
For Use With (Application) food processing, battery manufacturing, Paper pulp processing, sand blasting, pellet-blasting applications, fuel handling and refineries, masonry and concrete, hexavalent Chromium protection
Material Neoprene
Coating Material Tractor tread
Lining Flock