SHOWA™ 540 Cut Resistant Gloves
Offers a combination of exceptional grip, dexterity and cut-resistance with the durability of a Polyurethane coating
Manufacturer:  SHOWA™ 540M
 View more versions of this product
Catalog No. 191305613B
Description
Description
HPPE lining provides extremely comfortable fit
  • Durable Polyurethane palm-coated glove
  • A13-Gauge cut-resistant liner consisting of High Performance Polyethylene Engineered (HPPE)
  • Elastic knit-wrist secures the glove above the wrist for comfort and preventing debris from entering the glove
  • ANSICUT LEVEL 2†
  • Launders at 40°C (104°F) using a neutral detergent
†ANSI/IS1EA 105-2005 American National Standard for Hand Protection Selection Criteria
Specifications
Specifications
Polyurethane
Knit
HPPE (High-performance Polyethylene)
White
Level 2
machining, scrap Metal handling, glazing, Glass handling, automotive industry, cutting fluids, construction, HVAC, electronic assembly, maintenance
Medium
Documents