Corning™ Shorty Stripette™ Serological Pipets
Specially sized for small spaces
$155.51 - $295.15
Specifications
Sterile Yes
Length (Metric) 22cm
Material Polystyrene
Packaging Individually wrapped
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Packaging Quantity Price Quantity  
07-200-13 Corning™
C4103
10mL Case of 100 Case of 100 Each for $155.51
07-200-10 Corning™
C4102
5mL Case of 200 Case of 200 Each for $295.15
Specifications
Description
Shorter than standard-sized pipets, Shorty Stripettes move freely under the hood or in other confined spaces.
  • 32% shorter than standard Corning Stripette pipets
  • One-piece design reduces the possibility of particulates
  • Printed stripe magnifies the meniscus, enabling easier volume reading
  • Sterile, individually wrapped in paper/plastic
  • Nonpyrogenic
  • Noncytotoxic
Order Info
Individually wrapped in paper/plastic
Description & Specifications
Specifications
Yes
22cm
Polystyrene
Individually wrapped