Thermo Scientific™ Shelving Options and Accessories for Environmental Chambers and Incubators
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.
$495.33 - $797.50
Disclaimers:
Cat No: 224161 -- Two per unit max., not for shakers; 68.0 kg load maximum with shelf fully inserted and stationary
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
22-413-9 Thermo Scientific™
PSI224139
Solid Stainless Steel Shelf and Channels Each for $495.33
22-415-5 Thermo Scientific™
PSI224155
Perforated Stainless Steel Shelf and Channels Each for $725.79
22-416-1 Thermo Scientific™
PSI224161
Reinforced Stainless Steel Shelf and Channels (max. load 68kg) Each for $797.50
Specifications
Description
Utilize shelving options and accessories to increase the flexibility of your Thermo Scientific™ Environmental Chamber or Incubator.