Please login to your online account to display your discounted pricing

Sharp Point Forceps


Steel construction ensures years of service in every kind of dissecting work.

Manufacturer: fisher science education  0112CH

 View more versions of this product

Catalog No. S17313

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Length (English) 4.5 in.
Material Chrome Plated Steel
Length (Metric) 11.43cm
Tip Style Straight Sharp Point
Quantity (No. per Case) Each
Style Straight
Product Type Sharp Forceps with Corrugated Handles