Epredia™ Shandon™ Wright-Giemsa Stain Kit, Wright-Giemsa solution
Eosinates of polychrome methylene blue yield rich color
Manufacturer:  Epredia™ 9990710
 View more versions of this product
Catalog No. 28-613-5
Description
Description
  • Unique manufacturing process produces an intense differential stain for blood smears, bone marrows, mast cells, and FNA's
  • Demonstrates microorganisms such as Microfilaria, Babesia, Haemobartonella, Giardia, Pneumocystis, and Helicobacter
Specifications
Specifications
500 mL
Wright-Giemsa Stain Kit
Stain, buffer and rinse