Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
$126.76 - $1097.74
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
28-618-13
|
Epredia™
3120123 |
Multiple |
Each for $1,097.74
|
|
|||||
28-603-50
|
Epredia™
3120124 |
Blue |
Each for $128.71
|
|
|||||
28-603-56
|
Epredia™
3120125 |
Black |
Each for $128.85
|
|
|||||
29-603-57
|
Epredia™
3120126 |
Yellow |
Each for $146.47
|
|
|||||
28-603-58
|
Epredia™
3120127 |
Red |
Each for $126.76
|
|
|||||
28-603-62
|
Epredia™
3120128 |
Green |
Each for $129.21
|
|
|||||
Description
- Special binding pigment evenly coats the tissue without penetrating tissue surface
- Does not interfere with diagnostic interpretation
- Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Tissue Marking Dye |