Please login to your online account to display your discounted pricing

Thermo Scientific™ Shandon™ Halstead's Hemostatic Mosquito Straight Forceps

Choose 5″ (12.7cm) curved Halstead's Hemostatic Mosquito Straight Forceps for quality and precision.

$91.17 - $91.17

Specifications

Type Halstead
Length (English) 5 in.
Length (Metric) 12.7cm
Shape Straight
View More Specs
Products
Catalog Number Mfr. No. Grade Price Quantity    

38-106-60

 
thermo scientific™
SH507
Standard Each for $91.17
Description & Specifications

Specifications

Type Halstead
Length (English) 5 in.
Length (Metric) 12.7cm
Shape Straight

Choose 5″ (12.7cm) curved Thermo Scientific™ Shandon™ Halstead's Hemostatic Mosquito Straight Forceps for quality and precision. Forceps are available in premium and standard grades.

Features:

  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship